missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Cyclopentyl-3,5-dinitrobenzamide, 97%

Brand:  Alfa Aesar™ H59164.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15485388

  • £358.49 / 1g


Estimated Shipment date
from Supplier 10-05-2018
Add to basket

Specifications

Chemical Identifiers

N-Cyclopentyl-3,5-dinitrobenzamide
279.25
MFCD00707284

Specifications

N-Cyclopentyl-3,5-dinitrobenzamide
97%
MFCD00707284
Solid
279.25
1g
Videos
SDS