missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Cyclopentyl-3,4-difluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H61560.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15448128

  • £73.56 / 1g


Estimated Shipment date
from Supplier 30-01-2018
Add to basket

Specifications

Chemical Identifiers

N-Cyclopentyl-3,4-difluorobenzenesulfonamide
261.29
MFCD03975500

Specifications

N-Cyclopentyl-3,4-difluorobenzenesulfonamide
97%
MFCD03975500
Solid
261.29
1g
Videos
SDS