missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-CBZ-D-glutamic acid α-benzyl ester, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | 97°C to 98°C |
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (HPLC) |
Packaging | Glass Bottle |
Quantity | 1 g |
Specific Rotation | 24 |
Formula Weight | 371.39 |
Show More |
Safety and Handling
Product Identifier
- N-Cbz-D-Glutamic acid alpha-benzyl ester
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'