missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzyloxy-L-proline 99%, ACROS Organics™

Brand:  Acros Organics 108370050

Code : 29

Additional Details : CAS Number : 1148-11-4 Weight : 0.05500kg

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10667484

  • £21.40 / 5g


EU Stock 2
Add to basket

Specifications

Chemical Identifiers

N-Carbobenzyloxy-L-proline
1148-11-4
100.0
99%
249.27
N-CBZ-L-Proline
99%
98.5
≥98.5%
C13H15NO4
MFCD00003170

Specifications

N-Carbobenzyloxy-L-proline
White to off-white
99%
98.5
100.0
Crystalline Powder or Crystals
99%
249.27
Glass bottle
-59° to -61° (20°C, 589nm) (c=2, CH3COOH)
− 60.00 (20.00°C c=2,AcOH)
5g
1148-11-4
D-enantiomer: < 0.5%
Authentic
≥98.5%
C13H15NO4
MFCD00003170
N-CBZ-L-Proline
0.5% max. (K.F.)
Videos
SDS