missing translation for 'onlineSavingsMsg'
Learn More

N-ε-Carbobenzyloxy-L-lysine, 98%, Thermo Scientific Chemicals

Catalog Number 10676482
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
1155-64-2
C14H20N2O4
280.324
MFCD00002638
CKGCFBNYQJDIGS-LBPRGKRZSA-N
h-lys z-oh, n6-cbz-l-lysine, n-epsilon-carbobenzyloxy-l-lysine, n epsilon-benzyloxycarbonyl-l-lysine, n6-benzyloxycarbonyl-l-lysine, nepsilon-carbobenzoxy-l-lysine, nepsilon-carbobenzyloxy-l-lysine, l-lys cbz-oh, 2s-2-amino-6-benzyloxy carbonyl amino hexanoic acid, 6-n-cbz-l-lysine
1715626
C1=CC=C(C=C1)COC(=O)NCCCCC(C(=O)O)N
This item is not returnable. View return policy
1155-64-2
C14H20N2O4
280.324
MFCD00002638
CKGCFBNYQJDIGS-LBPRGKRZSA-N
h-lys z-oh, n6-cbz-l-lysine, n-epsilon-carbobenzyloxy-l-lysine, n epsilon-benzyloxycarbonyl-l-lysine, n6-benzyloxycarbonyl-l-lysine, nepsilon-carbobenzoxy-l-lysine, nepsilon-carbobenzyloxy-l-lysine, l-lys cbz-oh, 2s-2-amino-6-benzyloxy carbonyl amino hexanoic acid, 6-n-cbz-l-lysine
1715626
C1=CC=C(C=C1)COC(=O)NCCCCC(C(=O)O)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 259.0°C
CAS Min % 97.5
CAS Max % 100.0
Color White
Assay Percent Range 98%
Infrared Spectrum Authentic
Linear Formula C6H5CH2O2CNH(CH2)4CH(NH2)CO2H
Packaging Glass Bottle
Solubility Information Solubility in water: .. Other solubilities: soluble in most common organic solvents
IUPAC Name (2S)-2-amino-6-(phenylmethoxycarbonylamino)hexanoic acid
Specific Rotation + 15.50
Formula Weight 280.32
Percent Purity 98%
Sulfated Ash 0.2% max.
Water 0.5% max. (K.F.)
Quantity 5 g
Physical Form Powder
Chemical Name or Material Nε-Carbobenzyloxy-L-lysine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'