missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzyloxy-D-alanine, 98%, Thermo Scientific™

Catalog Number 11905571
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
26607-51-2
C11H13NO4
223.23
MFCD00063126
TYRGLVWXHJRKMT-UHFFFAOYNA-N
n-cbz-d-alanine, z-d-ala-oh, cbz-d-ala-oh, z-d-alanine, n-benzyloxycarbonyl-d-alanine, n-carbobenzoxy-d-alanine, r-2-benzyloxy carbonyl amino propanoic acid, n-carbobenzyloxy-d-alanine, benzyloxycarbonyl-d-alanine, cbz-d-alanine
70832
(2R)-2-(phenylmethoxycarbonylamino)propanoic acid
CC(NC(=O)OCC1=CC=CC=C1)C(O)=O
This item is not returnable. View return policy
26607-51-2
C11H13NO4
223.23
MFCD00063126
TYRGLVWXHJRKMT-UHFFFAOYNA-N
n-cbz-d-alanine, z-d-ala-oh, cbz-d-ala-oh, z-d-alanine, n-benzyloxycarbonyl-d-alanine, n-carbobenzoxy-d-alanine, r-2-benzyloxy carbonyl amino propanoic acid, n-carbobenzyloxy-d-alanine, benzyloxycarbonyl-d-alanine, cbz-d-alanine
70832
(2R)-2-(phenylmethoxycarbonylamino)propanoic acid
CC(NC(=O)OCC1=CC=CC=C1)C(O)=O
CAS Min % 97.5
CAS Max % 100.0
Melting Point 74°C to 80°C
Color White
Loss on Drying 1.00% max. ()
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Specific Rotation + 15.3
Formula Weight 223.23
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzyloxy-D-alanine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'