missing translation for 'onlineSavingsMsg'
Learn More

n-Butyric Anhydride, 98%, Thermo Scientific Chemicals

Catalog Number p-3735897
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
106-31-0
C8H14O3
158.20
MFCD00009389
YHASWHZGWUONAO-UHFFFAOYSA-N
butyric anhydride, n-butyric anhydride, butanoic anhydride, butanoic acid, anhydride, butyryl oxide, butyric acid anhydride, n-butyric acid anhydride, butyranhydrid, butanoic acid anhydride, butyric anhydride n
7798
butanoyl butanoate
CCCC(=O)OC(=O)CCC
This item is not returnable. View return policy
106-31-0
C8H14O3
158.20
MFCD00009389
YHASWHZGWUONAO-UHFFFAOYSA-N
butyric anhydride, n-butyric anhydride, butanoic anhydride, butanoic acid, anhydride, butyryl oxide, butyric acid anhydride, n-butyric acid anhydride, butyranhydrid, butanoic acid anhydride, butyric anhydride n
7798
butanoyl butanoate
CCCC(=O)OC(=O)CCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point -73.0°C
Color Colorless to Yellow
Density 0.9630g/mL
Boiling Point 195.0°C
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3CH2CH2CO)2O
Refractive Index 1.4120 to 1.4140
Quantity 100 mL
Beilstein 02, 274
Fieser 05,88
Merck Index 15, 1598
Specific Gravity 0.963
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in ether,decomposes in alcohol
Formula Weight 158.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material n-Butyric anhydride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'