missing translation for 'onlineSavingsMsg'
Learn More

n-Butyllithium, 2.6M solution in toluene, AcroSeal™, ACROS Organics™

Brand:  Acros Organics 378931000

Code : 29

Additional Details : CAS Number : 109-72-8 Weight : 0.13600kg Transport : UN number : 3394 Chem class : 4.2 Pack group : I


Quantity 100mL
Packaging AcroSeal™ Glass bottle

 View more versions of this product

Product Code. 10555183

  • £24.00 / 100mL


Call for latest availability
+44 (0)1509 555 500
Add to basket

This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
  Sea freight only: For EU stock allow up to 15 days delivery.

Specifications

Chemical Identifiers

n-Butyllithium
108-88-3
C4H9Li
64.06
2.6M solution in toluene
19.8 to 20.2% (as BuLi)
CH3(CH2)3Li
MFCD00009414

Specifications

n-Butyllithium
108-88-3
Solution
C4H9Li
64.06
MFCD00009414
0.4% max. (as BuLi)
Solubility in water: vigorous reaction.
Yellow to orange
4°C
2.6M solution in toluene
product may develop some turbidity or precipitate
19.8 to 20.2% (as BuLi)
CH3(CH2)3Li
0.8600g/mL
Solubility in water: vigorous reaction.
AcroSeal™ Glass bottle
0.86
100mL
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • n-Butyllithium, 2.6M solution in toluene

Signal Word
  • Danger

Hazard Category
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Pyrophoric liquid Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1

Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H336-May cause drowsiness or dizziness.
  • H250-Catches fire spontaneously if exposed to air.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P331-Do NOT induce vomiting.
  • P402+P404-Store in a dry place. Store in a closed container.

SDS