missing translation for 'onlineSavingsMsg'
Learn More

n-Butyltriphenylphosphonium bromide, 99%, Thermo Scientific Chemicals

Catalog Number p-80712
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1779-51-7
C22H24BrP
399.31
MFCD00011855
IKWKJIWDLVYZIY-UHFFFAOYSA-M
butyltriphenylphosphonium bromide, n-butyltriphenylphosphonium bromide, 1-butyl triphenylphosphonium bromide, phosphonium, butyltriphenyl-, bromide, butyl triphenyl phosphonium bromide, butyl triphenyl phosphine bromide, n-butyltriphenylphosphonium chloride, butyltriphenylphosphanium bromide, phosphonium, butyltriphenyl-, bromide 1:1, n-butyl triphenylphosphonium bromide
159628
[Br-].CCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
1779-51-7
C22H24BrP
399.31
MFCD00011855
IKWKJIWDLVYZIY-UHFFFAOYSA-M
butyltriphenylphosphonium bromide, n-butyltriphenylphosphonium bromide, 1-butyl triphenylphosphonium bromide, phosphonium, butyltriphenyl-, bromide, butyl triphenyl phosphonium bromide, butyl triphenyl phosphine bromide, n-butyltriphenylphosphonium chloride, butyltriphenylphosphanium bromide, phosphonium, butyltriphenyl-, bromide 1:1, n-butyl triphenylphosphonium bromide
159628
[Br-].CCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 240.0°C to 243.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula CH3(CH2)3P(C6H5)3Br
Quantity 5 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in chloroform,fairly soluble in lower alcohols,slightly soluble in aromatic hydrocarbons
IUPAC Name butyltriphenylphosphanium bromide
Formula Weight 399.3
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material n-Butyltriphenylphosphonium bromide, 99%
Show More Show Less
compliance-icons
Product Identifier
  • n-Butyltriphenylphosphonium bromide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'