missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl sulfide, 99%, pure, Thermo Scientific Chemicals

Catalog Number p-4509413
Click to view available options
Quantity:
5 mL
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
544-40-1
C8H18S
146.29
MFCD00009468
HTIRHQRTDBPHNZ-UHFFFAOYSA-N
dibutyl sulfide, butyl sulfide, n-butyl sulfide, butylthiobutane, di-n-butyl sulfide, butane, 1,1'-thiobis, butyl monosulfide, dibutyl thioether, thiononane-5, n-dibutyl sulfide
11002
CCCCSCCCC
This item is not returnable. View return policy
544-40-1
C8H18S
146.29
MFCD00009468
HTIRHQRTDBPHNZ-UHFFFAOYSA-N
dibutyl sulfide, butyl sulfide, n-butyl sulfide, butylthiobutane, di-n-butyl sulfide, butane, 1,1'-thiobis, butyl monosulfide, dibutyl thioether, thiononane-5, n-dibutyl sulfide
11002
CCCCSCCCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 0.8400g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula [CH3(CH2)3]2S
Refractive Index 1.4520 to 1.4540 (20°C, 589nm)
Quantity 5 mL
IUPAC Name 1-(butylsulfanyl)butane
Formula Weight 146.29
Percent Purity ≥98.5%
Grade Pure
Physical Form Liquid
Chemical Name or Material n-Butyl sulfide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'