missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl dodecanoate, 99+%, Thermo Scientific Chemicals

Catalog Number p-7051816
Click to view available options
Quantity:
25 mL
100 mL
500 mL
This item is not returnable. View return policy
106-18-3
C16H32O2
256.43
MFCD00042870
NDKYEUQMPZIGFN-UHFFFAOYSA-N
butyl laurate, dodecanoic acid, butyl ester, n-butyl laurate, bytyl laurate, butyl dodecylate, lauric acid butyl ester, lauric acid, butyl ester, lauric acid n-butyl ester, unii-23985ocm4h, n-butyl n-dodecanoate
61015
butyl dodecanoate
CCCCCCCCCCCC(=O)OCCCC
This item is not returnable. View return policy
106-18-3
C16H32O2
256.43
MFCD00042870
NDKYEUQMPZIGFN-UHFFFAOYSA-N
butyl laurate, dodecanoic acid, butyl ester, n-butyl laurate, bytyl laurate, butyl dodecylate, lauric acid butyl ester, lauric acid, butyl ester, lauric acid n-butyl ester, unii-23985ocm4h, n-butyl n-dodecanoate
61015
butyl dodecanoate
CCCCCCCCCCCC(=O)OCCCC

n-Butyl dodecanoate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -10°C
Density 0.86 g/mL
Boiling Point 166°C
Flash Point >110°C (230°F)
Refractive Index 1.435
Quantity 25 mL
Beilstein 1776360
Solubility Information Soluble in water,0.06006mg/L @ 25°C.
Formula Weight 256.4
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material n-Butyl dodecanoate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'