missing translation for 'onlineSavingsMsg'
Learn More

N-Butyl Acetate, Extra Pure, SLR, Fisher Chemical™

Brand:  Fisher Chemical B/4950/17

Code : 21

Additional Details : CAS Number : 123-86-4 Weight : 2.20500kg Transport : UN number : 1123 Chem class : 3 Pack group : III


 View more versions of this product

Product Code. 10090050

  • £106.00 / 2.50 L

In stock, ready to ship: 10

Please to purchase this item. Need a web account? Register with us today!


For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

Description

Description

N-Butyl Acetate, Extra Pure, SLR

Specifications

Specifications

Amber glass bottle
126°C
-90°C
6.2
C6H12O2
116.16g/mol
4
Colorless
2.5L
123-86-4
9445
Liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • n-Butyl acetate

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

SDS Documents
Certificates