missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Butyl acetate, ACS reagent, Thermo Scientific Chemicals
12.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
10.60 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "22914" to get your promotional price.
10.60 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -78.0°C |
Color | Colorless |
Density | 0.8800g/mL |
Boiling Point | 127.0°C |
Flash Point | 27°C |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Linear Formula | CH3CO2(CH2)3CH3 |
Refractive Index | 1.3930 to 1.3950 |
Quantity | 100 mL |
Show More |
Safety and Handling
Product Identifier
- n-Butyl acetate
- Warning
- Flammable liquid Category 3
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'