missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl acetate, 99.5%, for biochemistry, AcroSeal™, ACROS Organics™

Brand:  Acros Organics 447050025

Code : A0

Additional Details : CAS Number : 123-86-4 Weight : 2.20000kg Transport : UN number : 1123 Chem class : 3 Pack group : III

Quantity 2.5L

 View more versions of this product

Product Code. 12653590

  • £94.73 / 2.50 L

Estimated Shipment
07-05-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

For biochemistry
Specifications

Chemical Identifiers

123-86-4
116.16
n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle
CHEBI:31328
CCCCOC(=O)C
C6H12O2
DKPFZGUDAPQIHT-UHFFFAOYSA-N
31272
butyl acetate

Specifications

0.01 max. at 320nm, 0.01 max. at 400nm, 0.03 max. at 300nm, 0.05 max. at 275nm, 0.2 max. at 260nm, 0.5 max. at 256nm, 1 max. at 254nm
0.8800g/mL
1.3930 to 1.3950
0.69 mPa.s (25°C)
-78.0°C
123-86-4
C6H12O2
15,1537
Solubility in water: 7g/L (20°C)
CCCCOC(=O)C
116.16
CHEBI:31328
99.5%
n-Butyl acetate
27°C
0.88
127.0°C
2.5L
99.4% min. (GC)
CH3CO2(CH2)3CH3
n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle
DKPFZGUDAPQIHT-UHFFFAOYSA-N
butyl acetate
31272
116.16
Biochemistry
Videos
SDS Documents