missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Butyl 4-hydroxybenzoate, 99+%, Thermo Scientific Chemicals
40.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 g
1000 g
5000 g
Description
It is used as an preservative in cosmetics such as eye shadow, foundation, sunscreen, facial moisturizer and skin anti-aging treatment. It is also used in medication suspensions, and as a flavoring additive in food.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as an preservative in cosmetics such as eye shadow, foundation, sunscreen, facial moisturizer and skin anti-aging treatment. It is also used in medication suspensions, and as a flavoring additive in food.
Solubility
Soluble in DMSO, ethyl acetate, methanol.
Notes
Store in refrigerator. Store away from oxidizing agents.
It is used as an preservative in cosmetics such as eye shadow, foundation, sunscreen, facial moisturizer and skin anti-aging treatment. It is also used in medication suspensions, and as a flavoring additive in food.
Solubility
Soluble in DMSO, ethyl acetate, methanol.
Notes
Store in refrigerator. Store away from oxidizing agents.
Specifications
Melting Point | 67°C to 71°C |
Quantity | 250 g |
Beilstein | 1103741 |
Merck Index | 14,1584 |
Solubility Information | Soluble in DMSO,ethyl acetate,methanol. |
Formula Weight | 194.23 |
Percent Purity | ≥99% |
Chemical Name or Material | n-Butyl 4-hydroxybenzoate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'