missing translation for 'onlineSavingsMsg'
Learn More

n-Butanol (Clear, Colorless Liq.), Fisher BioReagents

Brand:  Fisher Bioreagents BP505-500

Code : 32

Additional Details : CAS Number : 71-36-3 Weight : 0.45500kg Transport : UN number : 1120 Chem class : 1 Pack group : III


Quantity 500mL
Packaging Glass Bottle

 View more versions of this product

Product Code. 10294763

  • £23.48 / 500mL


EU Stock 8
Add to basket


Specifications

Chemical Identifiers

n-Butanol
Liquid
≥99.8 %
CH3CH2CH2CH2OH
1-Butanol
71-36-3
≥99.8%
C4H10O
74.12

Specifications

n-Butanol
0.005% max.
Not Detected Detected/Not Detect.
≥99.8%
C4H10O
74.12
15, 1542
Pass Test
0.1% max.
6.7mbar at 20°C
2.95 mPa/s at 20°C
117.6°C
−89°C
71-36-3
Vapor Pressure: 6.7mbar at 20°C
Liquid
≥99.8 %
CH3CH2CH2CH2OH
1120
1-Butanol
0.810g/cm³
Glass Bottle
0.1% max.
Butanol
Colorless
500mL
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1-Butanol

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.

SDS Documentation