missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Bromosuccinimide, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
100 g
500 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent, C-X bond formation reagent
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | 175.0°C to 178.0°C |
Color | White to Yellow |
Flash Point | 180°C |
Loss on Drying | 1% max. |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5 to 101.5% (on dry substance) (Iodometry) |
Packaging | Glass bottle |
Molecular Formula | C4H4BrNO2 |
Show More |
Safety and Handling
Product Identifier
- N-Bromosuccinimide
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'