missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-sarcosine methyl ester, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
N-Boc-sarcosine methyl ester is involved in the preparation of N-[(R, R)-(E)-1-arylmethyl-3-(2-oxo-azepan-3-yl) carbamoyl] allyl-N-methyl-3, 5-bis(trifluoromethyl) benzamides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Boc-sarcosine methyl ester is involved in the preparation of N-[(R, R)-(E)-1-arylmethyl-3-(2-oxo-azepan-3-yl) carbamoyl] allyl-N-methyl-3, 5-bis(trifluoromethyl) benzamides.
Notes
Incompatible with oxidizing agents. Heat sensitive.
N-Boc-sarcosine methyl ester is involved in the preparation of N-[(R, R)-(E)-1-arylmethyl-3-(2-oxo-azepan-3-yl) carbamoyl] allyl-N-methyl-3, 5-bis(trifluoromethyl) benzamides.
Notes
Incompatible with oxidizing agents. Heat sensitive.
Specifications
Color | Yellow |
Linear Formula | (CH3)3COCON(CH3)CH2CO2CH3 |
IUPAC Name | methyl 2-{[(tert-butoxy)carbonyl](methyl)amino}acetate |
Formula Weight | 203.24 |
Percent Purity | 97% |
Quantity | 5 g |
Physical Form | Liquid |
Chemical Name or Material | N-Boc-sarcosine methyl ester |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'