missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-S-acetamidomethyl-D-cysteine, 97%

Brand:  Alfa Aesar™ H62978.06

Code : NEW

Additional Details : CAS Number : 138775-00-5 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15409418

  • £162.18 / 5g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

N-Boc-S-acetamidomethyl-D-cysteine
292.35
Boc-D-Cys(Acm)-OH; (S)-3-(Acetamidothio)-2-[(tert-butoxycarbonyl)(methyl)amino]propionic acid
138775-00-5
MFCD00076917

Specifications

N-Boc-S-acetamidomethyl-D-cysteine
Powder
292.35
Boc-D-Cys(Acm)-OH; (S)-3-(Acetamidothio)-2-[(tert-butoxycarbonyl)(methyl)amino]propionic acid
5g
138775-00-5
97%
MFCD00076917
White
Videos
SDS