missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-S-(4-methylbenzyl)-D-cysteine, 95%

Brand:  Alfa Aesar™ H62074.06

Code : NEW

Additional Details : CAS Number : 61925-78-8 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15468658

  • £135.68 / 5g


Estimated Shipment date
from Supplier 03-05-2018
Add to basket

Specifications

Chemical Identifiers

N-Boc-S-(4-methylbenzyl)-D-cysteine
325.42
Boc-D-Cys(4-MeBzl)-OH; (S)-2-(Boc-amino)-3-(4-methylbenzylthio)propionic acid
61925-78-8
MFCD00076924

Specifications

N-Boc-S-(4-methylbenzyl)-D-cysteine
Solid
325.42
Boc-D-Cys(4-MeBzl)-OH; (S)-2-(Boc-amino)-3-(4-methylbenzylthio)propionic acid
61925-78-8
95%
MFCD00076924
5g
Videos
SDS