missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-Piperidine-4-carbonitrile, 97%, Thermo Scientific™

Catalog Number p-4258794
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
91419-52-2
C11H18N2O2
210.28
UQADQTBQNVARAP-UHFFFAOYSA-N
1-boc-4-cyanopiperidine, 1-n-boc-4-cyanopiperidine, n-boc-4-cyanopiperidine, 1-n-boc-4-cyano-piperidine, n-boc-4-cyano-piperidine, 1-piperidinecarboxylic acid, 4-cyano-, 1,1-dimethylethyl ester, boc-4-cyanopiperidine, 4-cyano-piperidine-1-carboxylic acid tert-butyl ester, tert-butyl 4-cyano-1-piperidinecarboxylate, n-boc-piperidine-4-carbonitrile
1514443
tert-butyl 4-cyanopiperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(CC1)C#N
This item is not returnable. View return policy
91419-52-2
C11H18N2O2
210.28
UQADQTBQNVARAP-UHFFFAOYSA-N
1-boc-4-cyanopiperidine, 1-n-boc-4-cyanopiperidine, n-boc-4-cyanopiperidine, 1-n-boc-4-cyano-piperidine, n-boc-4-cyano-piperidine, 1-piperidinecarboxylic acid, 4-cyano-, 1,1-dimethylethyl ester, boc-4-cyanopiperidine, 4-cyano-piperidine-1-carboxylic acid tert-butyl ester, tert-butyl 4-cyano-1-piperidinecarboxylate, n-boc-piperidine-4-carbonitrile
1514443
tert-butyl 4-cyanopiperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(CC1)C#N
CAS Min % 96.0
CAS Max % 100.0
Melting Point 59°C to 63°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 210.28
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material N-BOC-Piperidine-4-carbonitrile
Show More Show Less
compliance-icons
Product Identifier
  • N-BOC-Piperidine-4-carbonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'