missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-2-Piperidinecarboxylic acid, 97+%, Acros Organics

£16.70 - £167.90

Chemical Identifiers

CAS 98303-20-9
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.28
MDL Number MFCD01862877
InChI Key JQAOHGMPAAWWQO-UHFFFAOYSA-N
Synonym n-boc-2-piperidinecarboxylic acid, 1-tert-butoxycarbonyl piperidine-2-carboxylic acid, 1-boc-piperidine-2-carboxylic acid, n-boc-dl-pipecolinic acid, n-boc-piperidine-2-carboxylic acid, boc-dl-pip-oh, n-boc-pipecolic acid, 1-n-boc-2-piperidinecarboxylic acid, 1-tert-butoxy carbonyl piperidine-2-carboxylic acid, 1-tert-butoxycarbonyl-2-piperidinecarboxylic acid
PubChem CID 581831
IUPAC Name 1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCCCC1C(=O)O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10107053 SDS View Documents Acros Organics
384760010
1g Glass bottle £16.70
1g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 05-02-2020
Please to purchase this item. Need a web account? Register with us today!
10014923 SDS View Documents Acros Organics
384760050
5g Glass bottle £26.70
5g View alternative products
Save Up to 
»
EU Stock 2 Please to purchase this item. Need a web account? Register with us today!
10433945 SDS View Documents Acros Organics
384760500
50g Glass bottle £167.90
50g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 05-02-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

98303-20-9
229.28
JQAOHGMPAAWWQO-UHFFFAOYSA-N
581831
CC(C)(C)OC(=O)N1CCCCC1C(=O)O
C11H19NO4
MFCD01862877
n-boc-2-piperidinecarboxylic acid, 1-tert-butoxycarbonyl piperidine-2-carboxylic acid, 1-boc-piperidine-2-carboxylic acid, n-boc-dl-pipecolinic acid, n-boc-piperidine-2-carboxylic acid, boc-dl-pip-oh, n-boc-pipecolic acid, 1-n-boc-2-piperidinecarboxylic acid, 1-tert-butoxy carbonyl piperidine-2-carboxylic acid, 1-tert-butoxycarbonyl-2-piperidinecarboxylic acid
1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-2-carboxylic acid

Specifications

N-BOC-2-Piperidinecarboxylic acid
Glass bottle
132°C to 134°C
98303-20-9
100.0
C11H19NO4
n-boc-2-piperidinecarboxylic acid, 1-tert-butoxycarbonyl piperidine-2-carboxylic acid, 1-boc-piperidine-2-carboxylic acid, n-boc-dl-pipecolinic acid, n-boc-piperidine-2-carboxylic acid, boc-dl-pip-oh, n-boc-pipecolic acid, 1-n-boc-2-piperidinecarboxylic acid, 1-tert-butoxy carbonyl piperidine-2-carboxylic acid, 1-tert-butoxycarbonyl-2-piperidinecarboxylic acid
CC(C)(C)OC(=O)N1CCCCC1C(=O)O
229.28
229.28
97+%
Authentic
White
1g
97.0
97+%
MFCD01862877
JQAOHGMPAAWWQO-UHFFFAOYSA-N
1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-2-carboxylic acid
581831
Crystalline Powder
Safety and Handling

Safety and Handling

Product Identifier
  • N-BOC-Piperidine-2-carboxylic acid

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.