missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-O-tert-butyl-D-threonine, 95%

Brand:  Alfa Aesar™ H62311.06

Code : NEW

Additional Details : CAS Number : 201217-86-9 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15438848

  • £315.88 / 5g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

N-Boc-O-tert-butyl-D-threonine
275.34
Boc-D-Thr(tBu)-OH; (2R,3S)-2-(Boc-amino)-3-(tert-butoxy)butyric acid
201217-86-9
MFCD00236876

Specifications

N-Boc-O-tert-butyl-D-threonine
Solid
275.34
Boc-D-Thr(tBu)-OH; (2R,3S)-2-(Boc-amino)-3-(tert-butoxy)butyric acid
201217-86-9
95%
MFCD00236876
5g
Videos
SDS