missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-O-benzyl-D-tyrosine, 95%

Brand:  Alfa Aesar™ H63427.06

Code : NEW

Additional Details : CAS Number : 63769-58-4 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15440119

  • £117.40 / 5g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Description

Description

Applications
N-Boc-O-benzyl-D-tyrosine is used as a intermediate for pharmaceutical and chemical research.

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications

Chemical Identifiers

N-Boc-O-benzyl-D-tyrosine
371.43
Boc-D-Tyr(Bzl)-OH; N-tert-Butoxycarbonyl-O-benzyl-D-tyrosine
63769-58-4
MFCD00038249

Specifications

N-Boc-O-benzyl-D-tyrosine
Solid
371.43
Boc-D-Tyr(Bzl)-OH; N-tert-Butoxycarbonyl-O-benzyl-D-tyrosine
63769-58-4
95%
MFCD00038249
5g
Videos
SDS