missing translation for 'onlineSavingsMsg'
Learn More

N-α-BOC-N-β-Xanthenyl-L-asparagine, 96%, Thermo Scientific™

Catalog Number p-4258791
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
65420-40-8
C22H24N2O6
412.44
MFCD00066157
YMGDQLXBNMRJMR-UHFFFAOYNA-N
boc-asn xan-oh, s-4-9h-xanthen-9-yl amino-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-alpha-t-boc-n-beta-xanthyl-l-asparagine, n-boc-n-xanthyl-l-asparagine, 2s-2-tert-butoxycarbonyl amino-3-9h-xanthen-9-yl carbamoyl propanoic acid, ambotzbaa1070, pubchem18926, nalpha-boc-nbeta-xanthenyl-l-asparagine, n alpha-boc-n gamma-9-xanthenyl-l-asparagine
7017974
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-(9H-xanthen-9-ylamino)butanoic acid
CC(C)(C)OC(=O)NC(CC(=O)NC1C2=CC=CC=C2OC2=CC=CC=C12)C(O)=O
This item is not returnable. View return policy
65420-40-8
C22H24N2O6
412.44
MFCD00066157
YMGDQLXBNMRJMR-UHFFFAOYNA-N
boc-asn xan-oh, s-4-9h-xanthen-9-yl amino-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-alpha-t-boc-n-beta-xanthyl-l-asparagine, n-boc-n-xanthyl-l-asparagine, 2s-2-tert-butoxycarbonyl amino-3-9h-xanthen-9-yl carbamoyl propanoic acid, ambotzbaa1070, pubchem18926, nalpha-boc-nbeta-xanthenyl-l-asparagine, n alpha-boc-n gamma-9-xanthenyl-l-asparagine
7017974
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-(9H-xanthen-9-ylamino)butanoic acid
CC(C)(C)OC(=O)NC(CC(=O)NC1C2=CC=CC=C2OC2=CC=CC=C12)C(O)=O
CAS Min % 95.0
CAS Max % 100.0
Melting Point 170°C to 172°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Specific Rotation 9.5
Formula Weight 412.45
Percent Purity 96%
Physical Form Powder
Chemical Name or Material N-α-BOC-N-β-Xanthenyl-L-asparagine
Show More Show Less
Product Identifier
  • N-alpha-BOC-N-beta-Xanthenyl-L-asparagine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'