missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-N-Methylethylenediamine, 97%, ACROS Organics™

Brand:  Acros Organics 437340050

Code : 29

Additional Details : CAS Number : 121492-06-6 Weight : 0.06000kg Transport : UN number : 2735 Chem class : 2b Pack group : III


Quantity 5mL
Packaging Glass bottle

 View more versions of this product

Product Code. 10169829

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

N-BOC-N-Methylethylenediamine
96.0
97%
C8H18N2O2
121492-06-6
100.0
97%
174.24

Specifications

N-BOC-N-Methylethylenediamine
96.0
100.0
Glass bottle
Liquid
97%
174.24
77.0° to 79.0°C (0.4 Torr)
>100°C
121492-06-6
0.9750g/mL
Authentic
0.975
97%
C8H18N2O2
Clear colorless to pale yellow
5mL
Videos
SDS