missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-N-methylethanolamine, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
N-Boc-N-methylethanolamine is an substituent in the synthesis of anticancer agent, an orally bioavailable small molecule inhibitor of signal transducer and activator of transcription.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Boc-N-methylethanolamine is an substituent in the synthesis of anticancer agent, an orally bioavailable small molecule inhibitor of signal transducer and activator of transcription.
Solubility
Soluble in diochloromethane. Slightly soluble in water.
Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
N-Boc-N-methylethanolamine is an substituent in the synthesis of anticancer agent, an orally bioavailable small molecule inhibitor of signal transducer and activator of transcription.
Solubility
Soluble in diochloromethane. Slightly soluble in water.
Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications
Color | Colorless to Yellow |
Beilstein | 3537964 |
Solubility Information | Soluble in diochloromethane. Slightly soluble in water. |
Formula Weight | 175.23 |
Percent Purity | 95% |
Quantity | 1 g |
Physical Form | Liquid |
Chemical Name or Material | N-Boc-N-methylethanolamine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'