missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-N-methyl-L-phenylglycine, 97%

Brand:  Alfa Aesar™ H63720.03

Code : NEW

Additional Details : CAS Number : 30925-11-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15420559

  • £36.70 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

N-Boc-N-methyl-L-phenylglycine
265.3
Boc-N-Me-Phg-OH
30925-11-2
MFCD00038799

Specifications

N-Boc-N-methyl-L-phenylglycine
Solid
265.3
Boc-N-Me-Phg-OH
30925-11-2
97%
MFCD00038799
1g
Videos
SDS