missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-N-methyl-L-phenylglycine, 97%, Thermo Scientific™

Catalog Number p-7055200
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
30925-11-2
C14H19NO4
265.31
MFCD00038799
COABPHLHHQAKPL-UHFFFAOYNA-N
boc-n-me-phg-oh, boc-n-methyl-l-phenylglycine, s-tert-butoxycarbonyl methyl amino phenyl acetic acid, s-2-tert-butoxycarbonyl methyl amino-2-phenylacetic acid, ambotzbaa1259, pubchem12248, boc-n-methyl-s-2-phenylglycine, s-methyl tert-butoxycarbonyl amino phenylacetic acid, n-alpha-t-butyloxycarbonyl-n-alpha-methyl-l-phenylglycine, 2s-tert-butoxycarbonyl methyl amino phenyl acetic acid
7010620
CN(C(C(O)=O)C1=CC=CC=C1)C(=O)OC(C)(C)C
This item is not returnable. View return policy
30925-11-2
C14H19NO4
265.31
MFCD00038799
COABPHLHHQAKPL-UHFFFAOYNA-N
boc-n-me-phg-oh, boc-n-methyl-l-phenylglycine, s-tert-butoxycarbonyl methyl amino phenyl acetic acid, s-2-tert-butoxycarbonyl methyl amino-2-phenylacetic acid, ambotzbaa1259, pubchem12248, boc-n-methyl-s-2-phenylglycine, s-methyl tert-butoxycarbonyl amino phenylacetic acid, n-alpha-t-butyloxycarbonyl-n-alpha-methyl-l-phenylglycine, 2s-tert-butoxycarbonyl methyl amino phenyl acetic acid
7010620
CN(C(C(O)=O)C1=CC=CC=C1)C(=O)OC(C)(C)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
IUPAC Name (2S)-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]-2-phenylacetic acid
Formula Weight 265.3
Percent Purity 97%
Quantity 250 mg
Physical Form Solid
Chemical Name or Material N-Boc-N-methyl-L-phenylglycine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'