missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-N-methyl-D-phenylalanine dicyclohexylammonium salt, 98%

Brand:  Alfa Aesar™ H62845.03

Code : NEW

Additional Details : CAS Number : 102185-45-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15487906

  • £59.78 / 1g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

N-Boc-N-methyl-D-phenylalanine dicyclohexylammonium salt
460.65
Boc-N-Me-D-Phe-OH.DCHA
102185-45-5
MFCD00058079

Specifications

N-Boc-N-methyl-D-phenylalanine dicyclohexylammonium salt
Solid
460.65
Boc-N-Me-D-Phe-OH.DCHA
102185-45-5
98%
MFCD00058079
1g
Videos
SDS