missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-N-methyl-D-alanine, 98%

Brand:  Alfa Aesar™ H62510.06

Code : NEW

Additional Details : CAS Number : 19914-38-6 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15449038

  • £397.50 / 5g


Estimated Shipment date
from Supplier 04-07-2018
Add to basket

Specifications

Chemical Identifiers

N-Boc-N-methyl-D-alanine
203.24
3606540
19914-38-6
MFCD00063137
Boc-N-Me-D-Ala-OH; N-tert-Butoxycarbonyl-N-methyl-D-alanine

Specifications

N-Boc-N-methyl-D-alanine
Powder
203.24
3606540
White
5g
19914-38-6
98%
MFCD00063137
Boc-N-Me-D-Ala-OH; N-tert-Butoxycarbonyl-N-methyl-D-alanine
90° to 94°C
Videos
SDS