missing translation for 'onlineSavingsMsg'
Learn More

Nα-BOC-L-Tryptophane, 97%, Thermo Scientific Chemicals

Catalog Number p-3776695
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
13139-14-5
C16H20N2O4
304.35
MFCD000655
NFVNYBJCJGKVQK-ZDUSSCGKSA-N
boc-trp-oh, n-boc-l-tryptophan, boc-l-tryptophan, n-tert-butoxy carbonyl-l-tryptophan, boc-l-trp-oh, boc-l-tryptophane, s-2-tert-butoxycarbonyl amino-3-1h-indol-3-yl propanoic acid, l-tryptophan, n-1,1-dimethylethoxy carbonyl, boc-tryptophan, n-tert-butoxycarbonyl-l-tryptophan
83169
CC(C)(C)OC(=O)N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O
This item is not returnable. View return policy
13139-14-5
C16H20N2O4
304.35
MFCD000655
NFVNYBJCJGKVQK-ZDUSSCGKSA-N
boc-trp-oh, n-boc-l-tryptophan, boc-l-tryptophan, n-tert-butoxy carbonyl-l-tryptophan, boc-l-trp-oh, boc-l-tryptophane, s-2-tert-butoxycarbonyl amino-3-1h-indol-3-yl propanoic acid, l-tryptophan, n-1,1-dimethylethoxy carbonyl, boc-tryptophan, n-tert-butoxycarbonyl-l-tryptophan
83169
CC(C)(C)OC(=O)N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color White
Assay Percent Range 97%
Infrared Spectrum Authentic
Beilstein 22, 6789
Packaging Glass Bottle
IUPAC Name (2S)-3-(1H-indol-3-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
Specific Rotation -19° to -23° (20°C, 589nm) (c=1, CH3COOH)
Formula Weight 304.34
Percent Purity ≥96.0%
Water 1% Max. (K.F.)
Quantity 5 g
Physical Form Powder
Chemical Name or Material Nα-BOC-L-Tryptophane
Afficher plus Afficher moins

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'