Alfa Aesar™ N-Boc-L-serine hydrate, 98%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H62458.06

UNSPSC: 12352209

Code: NEW

Additional Details:
Additional Details: CAS Number: 204191-40-2 Weight: 0.00500kg


Quantity 5g

Product Code. 15488968

Quantity Price
1 £ 149.46 / 5g
Estimated Shipment date
from Supplier 13-04-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material N-Boc-L-serine hydrate
CAS 204191-40-2
Formula Weight 223.22
MDL Number MFCD00150501
Synonym Boc-Ser-OH.H2O; (S)-2-(Boc-amino)-3-hydroxypropionic acid hydrate

Specification

Formula Weight 223.22
Physical Form Solid
Chemical Name or Material N-Boc-L-serine hydrate
Quantity 5g
Assay 98%
CAS 204191-40-2
Synonym Boc-Ser-OH.H2O; (S)-2-(Boc-amino)-3-hydroxypropionic acid hydrate
MDL Number MFCD00150501
Documentation

Technical documents