missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-L-proline methyl ester, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
N-Boc-L-proline methyl ester is generally used as a intermediate in pharmaceutical and chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Boc-L-proline methyl ester is generally used as a intermediate in pharmaceutical and chemical research.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
N-Boc-L-proline methyl ester is generally used as a intermediate in pharmaceutical and chemical research.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications
Quantity | 25 g |
Solubility Information | Slightly soluble in water. |
IUPAC Name | 1-O-tert-butyl 2-O-methyl (2S)-pyrrolidine-1,2-dicarboxylate |
Formula Weight | 229.27 |
Percent Purity | 96% |
Chemical Name or Material | N-Boc-L-proline methyl ester |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'