missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-L-Leucinol, 97%, Thermo Scientific™

Catalog Number p-3523511
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
82010-31-9
C11H23NO3
217.31
MFCD00076950
LQTMEOSBXTVYRM-VIFPVBQESA-N
boc-l-leucinol, n-boc-l-leucinol, boc-leucinol, boc-leu-ol, tert-butyl n-2s-1-hydroxy-4-methylpentan-2-yl carbamate, n-boc l-leucinol, pubchem12195, l-leucinol, n-boc protected, n-t-butoxycarbonyl-l-leucinol
7018766
tert-butyl N-[(2S)-1-hydroxy-4-methylpentan-2-yl]carbamate
CC(C)C[C@@H](CO)NC(=O)OC(C)(C)C
This item is not returnable. View return policy
82010-31-9
C11H23NO3
217.31
MFCD00076950
LQTMEOSBXTVYRM-VIFPVBQESA-N
boc-l-leucinol, n-boc-l-leucinol, boc-leucinol, boc-leu-ol, tert-butyl n-2s-1-hydroxy-4-methylpentan-2-yl carbamate, n-boc l-leucinol, pubchem12195, l-leucinol, n-boc protected, n-t-butoxycarbonyl-l-leucinol
7018766
tert-butyl N-[(2S)-1-hydroxy-4-methylpentan-2-yl]carbamate
CC(C)C[C@@H](CO)NC(=O)OC(C)(C)C

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Infrared Spectrum Authentic
Color Colorless to Yellow
Packaging Glass bottle
CAS Min % 96.0
Refractive Index 1.4470 to 1.4490 (20°C, 589nm)
CAS Max % 100.0
Specific Rotation -25° to -29° (20°C, 589nm) (C=2, methanol)
Quantity 1 g
Physical Form Viscous Liquid
Assay Percent Range 97%
Formula Weight 217.31
Percent Purity ≥96.0%
Chemical Name or Material N-BOC-L-Leucinol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'