missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-L-leucine methyl ester, 97%

Brand:  Alfa Aesar™ H61551.06

Code : NEW

Additional Details : CAS Number : 63096-02-6 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15488108

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

N-Boc-L-leucine methyl ester
245.32
Boc-Leu-OMe
63096-02-6
MFCD00076952

Specifications

N-Boc-L-leucine methyl ester
Liquid
245.32
1.4400
0.991g/mL
5g
63096-02-6
97%
MFCD00076952
Boc-Leu-OMe
205°C
Videos
SDS