missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-L-glutamine 4-nitrophenyl ester, 95%

Brand:  Alfa Aesar™ H62707.14

Code : NEW

Additional Details : CAS Number : 15387-45-8 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15499188

  • £54.27 / 25g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

N-Boc-L-glutamine 4-nitrophenyl ester
367.35
1896303
15387-45-8
MFCD00065572
Boc-Gln-ONp

Specifications

N-Boc-L-glutamine 4-nitrophenyl ester
Powder
367.35
1896303
White
Light Sensitive
15387-45-8
95%
MFCD00065572
Boc-Gln-ONp
25g
Videos
SDS