missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-L-glutamic acid 5-(9-fluorenylmethyl) ester, 98%

Brand:  Alfa Aesar™ H63101.06

Code : NEW

Additional Details : CAS Number : 123417-18-5 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15418126

  • £167.48 / 5g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

N-Boc-L-glutamic acid 5-(9-fluorenylmethyl) ester
425.47
Boc-Glu(Ofm)-OH
123417-18-5
MFCD00076934

Specifications

N-Boc-L-glutamic acid 5-(9-fluorenylmethyl) ester
Solid
425.47
Boc-Glu(Ofm)-OH
123417-18-5
98%
MFCD00076934
5g
Videos
SDS