missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-L-Aspartic acid, 99%, ACROS Organics™

Brand:  Acros Organics 303210010

Code : 29

Additional Details : CAS Number : 13726-67-5 Weight : 0.05100kg

Packaging Glass bottle
Quantity 1g

 View more versions of this product

Product Code. 11955471

  • £12.90 / 1g


EU Stock 2
Add to basket

Specifications

Chemical Identifiers

N-BOC-L-Aspartic acid
99%
HO2CCH2CH[NHCO2C(CH3)3]CO2H
N-(tert-Butoxycarbonyl)-L-aspartic acid
99%
C9H15NO6
233.22

Specifications

N-BOC-L-Aspartic acid
Authentic
99%
C9H15NO6
HO2CCH2CH[NHCO2C(CH3)3]CO2H
Solubility in water: slightly soluble in water.
− 6.20
116.0° to 120.0°C
1g
13726-67-5
Crystalline Powder
99%
Glass bottle
233.22
N-(tert-Butoxycarbonyl)-L-aspartic acid
0.5% Max. (K.F.)
White to off-white
Videos
SDS