missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-BOC-L-Aspartic acid, 99%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Specifications
Melting Point | 116.0°C to 120.0°C |
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Glass bottle |
Linear Formula | HO2CCH2CH[NHCO2C(CH3)3]CO2H |
Quantity | 5 g |
Specific Rotation | − 6.20 |
Solubility Information | Solubility in water: slightly soluble in water. |
Formula Weight | 233.22 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'