missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-L-aspartic acid 4-tert-butyl ester dicyclohexylammonium salt, 95%

Brand:  Alfa Aesar™ H61621.06

Code : NEW

Additional Details : CAS Number : 1913-12-8 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15456656

  • £19.82 / 5g


Estimated Shipment date
from Supplier 03-05-2018
Add to basket

Specifications

Chemical Identifiers

N-Boc-L-aspartic acid 4-tert-butyl ester dicyclohexylammonium salt
470.64
Boc-Asp(OtBu)-OH.DCHA
1913-12-8
MFCD00038889

Specifications

N-Boc-L-aspartic acid 4-tert-butyl ester dicyclohexylammonium salt
Solid
470.64
Boc-Asp(OtBu)-OH.DCHA
1913-12-8
95%
MFCD00038889
5g
Videos
SDS