missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-aspartic acid 4-tert-butyl ester, 98%, Thermo Scientific Chemicals

Catalog Number 15430309
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
1676-90-0
C13H23NO6
289.328
MFCD00076912
PHJDCONJXLIIPW-QMMMGPOBSA-N
boc-asp otbu-oh, boc-l-aspartic acid 4-tert-butyl ester, boc-asp obut-oh, s-4-tert-butoxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-alpha-t-boc-l-aspartic acid beta-t-butyl ester, 2s-4-tert-butoxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-asp but, pubchem12101, n-boc-asp o-t-bu oh
7010636
(2S)-4-[(2-methylpropan-2-yl)oxy]-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
CC(C)(C)OC(=O)CC(C(=O)O)NC(=O)OC(C)(C)C
This item is not returnable. View return policy
1676-90-0
C13H23NO6
289.328
MFCD00076912
PHJDCONJXLIIPW-QMMMGPOBSA-N
boc-asp otbu-oh, boc-l-aspartic acid 4-tert-butyl ester, boc-asp obut-oh, s-4-tert-butoxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-alpha-t-boc-l-aspartic acid beta-t-butyl ester, 2s-4-tert-butoxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-asp but, pubchem12101, n-boc-asp o-t-bu oh
7010636
(2S)-4-[(2-methylpropan-2-yl)oxy]-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
CC(C)(C)OC(=O)CC(C(=O)O)NC(=O)OC(C)(C)C

It is used in the preparation of highly selective thrombin inhibitors. Also used in the preparation of thiopeptides and thioacylating agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the preparation of highly selective thrombin inhibitors. Also used in the preparation of thiopeptides and thioacylating agents.

Solubility
Soluble in methanol and dimethyl sulfoxide.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 64°C to 67°C
Color White
Beilstein 2336144
Solubility Information Soluble in methanol and dimethyl sulfoxide.
Formula Weight 289.32
Percent Purity 98%
Quantity 5 g
Physical Form Powder
Chemical Name or Material N-Boc-L-aspartic acid 4-tert-butyl ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'