missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-L-aspartic acid 4-tert-butyl ester, 98%, Thermo Scientific Chemicals
25.73 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
It is used in the preparation of highly selective thrombin inhibitors. Also used in the preparation of thiopeptides and thioacylating agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the preparation of highly selective thrombin inhibitors. Also used in the preparation of thiopeptides and thioacylating agents.
Solubility
Soluble in methanol and dimethyl sulfoxide.
Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
It is used in the preparation of highly selective thrombin inhibitors. Also used in the preparation of thiopeptides and thioacylating agents.
Solubility
Soluble in methanol and dimethyl sulfoxide.
Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Melting Point | 64°C to 67°C |
Color | White |
Beilstein | 2336144 |
Solubility Information | Soluble in methanol and dimethyl sulfoxide. |
Formula Weight | 289.32 |
Percent Purity | 98% |
Quantity | 5 g |
Physical Form | Powder |
Chemical Name or Material | N-Boc-L-aspartic acid 4-tert-butyl ester |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'