missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-L-aspartic acid 4-cyclohexyl ester, 95%

Brand:  Alfa Aesar™ H61240.22

Code : NEW

Additional Details : CAS Number : 73821-95-1 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 15467748

  • £245.00 / 100g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

N-Boc-L-aspartic acid 4-cyclohexyl ester
315.36
3563575
73821-95-1
MFCD00061996
Boc-Asp(OcHx)-OH

Specifications

N-Boc-L-aspartic acid 4-cyclohexyl ester
Powder
315.36
3563575
White
100g
73821-95-1
95%
MFCD00061996
Boc-Asp(OcHx)-OH
93° to 95°C
Videos
SDS