missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-L-aspartic acid 4-benzyl 1-(4-nitrophenyl) ester, 98%

Brand:  Alfa Aesar™ H62895.06

Code : NEW

Additional Details : CAS Number : 26048-69-1 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15487966

  • £17.81 / 5g


Estimated Shipment date
from Supplier 07-02-2018
Add to basket

Specifications

Chemical Identifiers

N-Boc-L-aspartic acid 4-benzyl 1-(4-nitrophenyl) ester
C22H24N2O8
MFCD00038122
26048-69-1
444.43
Boc-Asp(OBzl)-ONp; (S)-4-Benzyl 2-(Boc-amino)-1-(4-nitrophenyl)succinate

Specifications

N-Boc-L-aspartic acid 4-benzyl 1-(4-nitrophenyl) ester
98%
444.43
5g
26048-69-1
C22H24N2O8
MFCD00038122
Boc-Asp(OBzl)-ONp; (S)-4-Benzyl 2-(Boc-amino)-1-(4-nitrophenyl)succinate
Videos
SDS