missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-L-aspartic acid 1-methyl ester, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
N-Boc-L-aspartic acid 1-methyl ester is used as an intermediate in the preparation of modified alfa-amino acids and 5-cis-alkylprolines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Boc-L-aspartic acid 1-methyl ester is used as an intermediate in the preparation of modified alfa-amino acids and 5-cis-alkylprolines.
Notes
Incompatible with oxidizing agents. Heat sensitive.
N-Boc-L-aspartic acid 1-methyl ester is used as an intermediate in the preparation of modified alfa-amino acids and 5-cis-alkylprolines.
Notes
Incompatible with oxidizing agents. Heat sensitive.
Specifications
Formula Weight | 247.25 |
Percent Purity | 98% |
Quantity | 250 mg |
Physical Form | Solid |
Chemical Name or Material | N-Boc-L-aspartic acid 1-methyl ester |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'