missing translation for 'onlineSavingsMsg'
Learn More

N-α-BOC-L-Asparagine, 99%, Thermo Scientific Chemicals

Catalog Number 11935471
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
7536-55-2
C9H16N2O5
232.24
FYYSQDHBALBGHX-YFKPBYRVSA-N
boc-asn-oh, boc-l-asparagine, boc-asn, n-tert-butoxycarbonyl-l-asparagine, tert-butoxycarbonylasparagine, nalpha-tert-butoxycarbonyl-l-asparagine, boc-asparagine, tert-butoxycarbonyl-l-asparagine, n-tert-butoxycarbonyl asparagine, tert-butyloxycarbonyl-l-asparagine
82035
CHEBI:3146
(2S)-4-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
CC(C)(C)OC(=O)NC(CC(=O)N)C(=O)O
This item is not returnable. View return policy
7536-55-2
C9H16N2O5
232.24
FYYSQDHBALBGHX-YFKPBYRVSA-N
boc-asn-oh, boc-l-asparagine, boc-asn, n-tert-butoxycarbonyl-l-asparagine, tert-butoxycarbonylasparagine, nalpha-tert-butoxycarbonyl-l-asparagine, boc-asparagine, tert-butoxycarbonyl-l-asparagine, n-tert-butoxycarbonyl asparagine, tert-butyloxycarbonyl-l-asparagine
82035
CHEBI:3146
(2S)-4-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
CC(C)(C)OC(=O)NC(CC(=O)N)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 170.0°C to 175.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula NH2COCH2CH(COOH)NHCOOC(CH3)3
Quantity 5 g
Specific Rotation − 7.90
Formula Weight 232.24
Percent Purity 99%
Physical Form Crystalline Powder or Powder
Chemical Name or Material N-α-BOC-L-Asparagine
Show More Show Less
compliance-icons
Product Identifier
  • N-alpha-BOC-L-Asparagine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'