missing translation for 'onlineSavingsMsg'
Learn More

N-α-BOC-L-Asparagine 4-nitrophenyl ester, 98%, Thermo Scientific™

Catalog Number p-4258782
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
4587-33-1
C15H19N3O7
353.33
IAPXDJMULQXGDD-NSHDSACASA-N
boc-asn-onp, boc-l-asparagine 4-nitrophenyl ester, boc-asn-ophno2, s-4-nitrophenyl 4-amino-2-tert-butoxycarbonyl amino-4-oxobutanoate, 4-nitrophenyl 2s-2-tert-butoxycarbonyl amino-3-carbamoylpropanoate, pubchem12095, threonin-tert.-butylether, ch3 3coconhch cooh ch2cooh, n1-t-boc c1-p-nitrophenol asparagine, boc-asn-onp hplc
439663
CHEBI:3147
(4-nitrophenyl) (2S)-4-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoate
CC(C)(C)OC(=O)NC(CC(=O)N)C(=O)OC1=CC=C(C=C1)[N+](=O)[O-]
This item is not returnable. View return policy
4587-33-1
C15H19N3O7
353.33
IAPXDJMULQXGDD-NSHDSACASA-N
boc-asn-onp, boc-l-asparagine 4-nitrophenyl ester, boc-asn-ophno2, s-4-nitrophenyl 4-amino-2-tert-butoxycarbonyl amino-4-oxobutanoate, 4-nitrophenyl 2s-2-tert-butoxycarbonyl amino-3-carbamoylpropanoate, pubchem12095, threonin-tert.-butylether, ch3 3coconhch cooh ch2cooh, n1-t-boc c1-p-nitrophenol asparagine, boc-asn-onp hplc
439663
CHEBI:3147
(4-nitrophenyl) (2S)-4-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoate
CC(C)(C)OC(=O)NC(CC(=O)N)C(=O)OC1=CC=C(C=C1)[N+](=O)[O-]
Melting Point 136°C to 140°C
CAS Min % 97.5
CAS Max % 100.0
Color White
Assay Percent Range 97.5% min. (HPLC)
Infrared Spectrum Authentic
Packaging Glass Bottle
Specific Rotation -36.4
Formula Weight 353.33
Percent Purity 98%
Water 1% Max.
Quantity 1 g
Physical Form Powder
Chemical Name or Material N-α-BOC-L-Asparagine 4-nitrophenyl ester
Show More Show Less
Product Identifier
  • N-alpha-BOC-L-Asparagine 4-nitrophenyl ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'