missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-guanidine, 95%

Brand:  Alfa Aesar™ H63206.03

Code : NEW

Additional Details : CAS Number : 219511-71-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15439778

  • £9.90 / 1g


Estimated Shipment date
from Supplier 04-05-2018
Add to basket

Description

Description

Applications
N-Boc-guanidine, is used as used as guanidinylating agent and for synthesis of 2-aminoimidazoles. It is also used as a pharmaceutical intermediate.

Solubility
Soluble in most alcohols, esters (e.g., ethyl acetate), ketones (e.g., acetone), acetonitrile, and dichloromethane. Not soluble in diethyl ether and hexane.

Notes
Moisture, heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

N-Boc-guanidine
(CH3)3OCONHC(=NH)NH2
MFCD20547465
219511-71-4
159.19

Specifications

N-Boc-guanidine
Moisture Sensitive
95%
159.19
MFCD20547465
1g
219511-71-4
Powder
(CH3)3OCONHC(=NH)NH2
170° to 175°C
White
Videos
SDS