missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-guanidine, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
N-Boc-guanidine, is used as used as guanidinylating agent and for synthesis of 2-aminoimidazoles. It is also used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Boc-guanidine, is used as used as guanidinylating agent and for synthesis of 2-aminoimidazoles. It is also used as a pharmaceutical intermediate.
Solubility
Soluble in most alcohols, esters (e.g., ethyl acetate), ketones (e.g., acetone), acetonitrile, and dichloromethane. Not soluble in diethyl ether and hexane.
Notes
Moisture, heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
N-Boc-guanidine, is used as used as guanidinylating agent and for synthesis of 2-aminoimidazoles. It is also used as a pharmaceutical intermediate.
Solubility
Soluble in most alcohols, esters (e.g., ethyl acetate), ketones (e.g., acetone), acetonitrile, and dichloromethane. Not soluble in diethyl ether and hexane.
Notes
Moisture, heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
Melting Point | 170°C to 175°C |
Color | White |
Linear Formula | (CH3)3OCONHC(=NH)NH2 |
Quantity | 5 g |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in most alcohols,esters (e.g.,ethyl acetate),ketones (e.g.,acetone),acetonitrile,and dichloromethane. Not soluble in diethyl ether and hexane. |
Formula Weight | 159.19 |
Percent Purity | 95% |
Physical Form | Powder |
Chemical Name or Material | N-Boc-guanidine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'